Learn More
Sartorius Cellulose Nitrate Membrane Filters
Product Code. p-4301121 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric):
13 mm
25 mm
47 mm
50 mm
90 mm
100 mm
Pore Size:
0.45 μm
0.65 μm
0.8 μm
1.2 μm
3 μm
5 μm
8 μm
Flow Rate:
130mL/min.
200mL/min.
430mL/min.
570mL/min.
70mL/min.
750mL/min.
Quantity:
100/Pk.
25/Pk.
Unit Size:
Pack of 100
Pack of 25
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Pore Size Flow Rate Quantity unitSize
10303591 13 mm 8 μm 750mL/min. 100/Pk. Pack of 100
10272221 47 mm 8 μm 750mL/min. 100/Pk. Pack of 100
10068070 100 mm 1.2 μm 200mL/min. 25/Pk. Pack of 25
10675051 50 mm 8 μm 750mL/min. 100/Pk. Pack of 100
10383441 25 mm 1.2 μm 200mL/min. 100/Pk. Pack of 100
10634751 47 mm 0.8 μm 200mL/min. 100/Pk. Pack of 100
10231651 47 mm 0.65 μm 130mL/min. 100/Pk. Pack of 100
10262131 25 mm 0.8 μm 200mL/min. 100/Pk. Pack of 100
10494261 25 mm 0.65 μm 130mL/min. 100/Pk. Pack of 100
10333491 100 mm 0.45 μm 70mL/min. 25/Pk. Pack of 25
10193891 90 mm 0.45 μm 70mL/min. 100/Pk. Pack of 100
10567711 25 mm 0.45 μm 70mL/min. 100/Pk. Pack of 100
10425401 50 mm 0.45 μm 70mL/min. 100/Pk. Pack of 100
10018070 47 mm 0.45 μm 70mL/min. 100/Pk. Pack of 100
12972661 47 mm 0.45 μm 70mL/min. 100/Pk. Pack of 100
10008060 90 mm 0.45 μm 70mL/min. 25/Pk. Pack of 25
10199130 25 mm 5 μm 570mL/min. 100/Pk. Pack of 100
10508871 50 mm 0.8 μm 200mL/min. 100/Pk. Pack of 100
10404651 25 mm 3 μm 430mL/min. 100/Pk. Pack of 100
10211751 47 mm 3 μm 430mL/min. 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Cellulose nitrate filters are highly flammable. Their flashpoint is approx. 200°C. Always keep these filters separated by the discs of interleaving paper that are supplied with the filters.
Product Code. 10303591 Supplier Sartorius Supplier No. 1130113N
Cellulose nitrate filters are highly flammable. Their flashpoint is approx. 200°C. Always keep these filters separated by the discs of interleaving paper that are supplied with the filters.
Combines uniform pore structure with high flow and adsorption rates. Sartorius Cellulose Nitrate Membrane Filters are excellently suited for diagnostic kits. Non-sterile, hydrophilic filters in a variety of diameters and pore sizes are bulk-packed to save money and space.
  • Cellulose nitrate filters combine high flow rates with high non-specific adsorption
  • Defined particle retention and outstanding, reliable capture rates for microorganisms
  • Very advantageous for diagnostic kits and compatible with aqueous solutions, hydrocarbons and several other organic solvents
  • Filters have a uniform pore structure and are available in range of diameters and pore sizes
    • Pore sizes: 0.45μm, 0.65μm, 0.8μm, 1.2μm, 3.0μm, 5μm and 8μm
    • The larger pore sizes (8 μm, 5 μm, 3 μm) are useful for cell retention and prefiltering
    • The 0.45 pore size is ideal for particle collection and filtration
    • Diameters: 13, 25, 37, 47, 50, 90 and 100mm (0.5 to 3.9 in.)
  • Ungridded filters can be sterilized via autoclave (wet) to 121°C, ethylene oxide or γ-irradiation to 25 kGy
  • Box label specifies the type, quantity, diameter, pore size and lot number
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Bubble Point 0.3bar
Color White
Disposable Single-use
Filter Type Membrane Filter
Format Disc
Material Cellulose Nitrate
Material (Membrane) Cellulose Nitrate
Pore Size 8 μm
Sterility Non-sterile
Flow Rate 750mL/min.
Gas Flow Rate 41L/min.
Hydrophobicity Hydrophilic
Quantity 100/Pk.
Thickness (Metric) 130 μm
Type Membrane Filter
Diameter (Metric) 13 mm
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P230-Keep wetted with .
  • P233-Keep container tightly closed.
  • P234-Keep only in original container.
  • P243-Take precautionary measures against static discharge.
  • P370+P378-In case of fire: Use to extinguish.
  • P372-Explosion risk in case of fire.
  • P375-Fight fire remotely due to the risk of explosion.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.