Description
Vivapure Mini Ion Exchange spin columns are centrifugal devices, based on Sartobind Membrane Adsorber technology as their chromatography matrix. The devices are ready-to-use and cannot run dry by their design. For many protein purification applications, they can replace time-consuming column chromatography. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many samples are processed parallel. Vivapure Mini is available with three different membrane chemistries and with medium (1mg/unit) or high (4mg/unit) protein binding capacity. Vivapure Mini units are intended for use in swing-out rotors and accommodate 0.5mL and 0.4mL samples, respectively.
- Fast centrifugal spin columns
- Easy and ready-to-use
- Binding capacity 1mg up to 4mg protein
- Ideal for parallel processing
Specifications
Specifications
| Bed Volume | 240 μL |
| Capacity (Metric) | 0.4 mL |
| Material | Polypropylene |
| Pore Size | 3 to 5 μm |
| Support Type | Stabilized regenerated Cellulose |
| Quantity | 16 Pack |
| For Use With (Application) | Sample fractionation, purification condition scouting, small scale purification |
| Type | Spin Column |