missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SATB2 Antibody (CL0323) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52916B
This item is not returnable.
View return policy
Description
SATB2 Monoclonal antibody specifically detects SATB2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| SATB2 | |
| Monoclonal | |
| Biotin | |
| DNA-binding protein SATB2, FLJ32076, KIAA1034FLJ21474, MGC119474, MGC119477, SATB family member 2, SATB homeobox 2, Special AT-rich sequence-binding protein 2 | |
| This antibody was developed using a recombinant protein derived from Q9UPW6, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Signal Transduction | |
| 23314 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL0323 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction