missing translation for 'onlineSavingsMsg'
Learn More

Saxagliptin, TRC

Product Code. 30006367
Change view
Click to view available options
Quantity:
1 mg
10 mg
100 mg
Unit Size:
100mg
10mg
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006367 10 mg 10mg
30134771 100 mg 100mg
17968884 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006367 Supplier Toronto Research Chemicals Supplier No. TRCS14350010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 361442-04-8
Molecular Formula C18 H25 N3 O2
Molecular Weight (g/mol) 315.41
Synonym (1S,3S,5S)-2-[(2S)-2-amino-2-(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-2-Azabicyclo[3.1.0]hexane-3-carbonitrile, 2-Azabicyclo[3.1.0]hexane-3-carbonitrile, 2-[(2S)-amino(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-, (1S,3S,5S)- (9CI), (1S,3S,5S)-2-[(2S)-2-Amino-2-(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile, BMS 477118, BMS 477118-11, Onglyza, Saxagliptin
IUPAC Name (1S,3S,5S)-2-[(2S)-2-amino-2-(3-hydroxy-1-adamantyl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile
SMILES N[C@H](C(=O)N1[C@@H](C[C@@H]2C[C@H]12)C#N)C34CC5CC(CC(O)(C5)C3)C4

Specifications

InChI Formula InChI=1S/C18H25N3O2/c19-8-13-2-12-3-14(12)21(13)16(22)15(20)17-4-10-1-11(5-17)7-18(23,6-10)9-17/h10-15,23H,1-7,9,20H2/t10?,11?,12-,13+,14+,15-,17?,18?/m1/s1
Quantity 10 mg
Formula Weight 315.1947
Percent Purity >95%
Shipping Condition Blue Ice
Chemical Name or Material Saxagliptin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.