missing translation for 'onlineSavingsMsg'
Learn More

SBDS Mouse anti-Human, Unconjugated, Clone: 1F8A10, Proteintech

Mouse Monoclonal Antibody

145.00€ - 410.00€

Specifications

Antigen SBDS
Clone 1F8A10
Concentration 1000 μg/mL
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16889705
View Documents
Proteintech
67200-1-IG-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16879705
View Documents
Proteintech
67200-1-IG-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SBDS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SBDS. This antibody recognizes human, mouse, rat antigen. The SBDS antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.

Specifications

Specifications

SBDS
1000 μg/mL
Monoclonal
Mouse
51119
Primary
1F8A10
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Human, Rat, Mouse
IgG2a
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.