Learn More
SCAMP2 Monoclonal Antibody (OTI2F1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA506992S
Encompass_Preferred
 View more versions of this product
Product Code. 17516712
  • 242.00€ / 30µL
Description
SCAMP2 Monoclonal antibody specifically detects SCAMP2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
SCAMP2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O15127
Mouse
Affinity chromatography
RUO
10066
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
OTI2F1
Unconjugated
Scamp2
AI875466; I79_023583; Sc2; scamp 2; SCAMP2; Secretory carrier membrane protein 2; secretory carrier-associated membrane protein 2; testis secretory sperm-binding protein Li 219p
Full length human recombinant protein of human SCAMP2 produced in HEK293T cell.