missing translation for 'onlineSavingsMsg'
Learn More

Scandium Std - Sc @ 5000μg/g in 75 cSt Oil

Product Code. 30314911
Change view
Click to view available options
Quantity:
200 g
400 g
Unit Size:
200g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314911 400 g -
30323884 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30314911 Supplier VHG Labs Supplier No. VHGOSC5000400G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Especificaciones

Chemical Name or Material Scandium
CAS 7440-20-2
Analyte or Component Names Scandium
Shipping Condition Room Temperature
Molecular Formula Sc
Concentration 5000 μg/g
Synonym Sc
InChI Formula InChI=1 S/Sc
Solution Type Single Solution
SMILES [Sc]
Element Sc
IUPAC Name scandium
Solvent or Matrix 75 cSt Oil
Industry Type Industrial QC
Molecular Weight (g/mol) 44.96
Quantity 400 g
Formula Weight 44.956
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.