missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCC112 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98627CL1
This item is not returnable.
View return policy
Description
SCC112 Polyclonal antibody specifically detects SCC112 in Human samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| SCC112 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| Cell proliferation-inducing gene 54 protein, DKFZp686B19246, FLJ41012, KIAA0648MGC131948, PDS5, PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae), PIG54, SCC-112MGC161503, Sister chromatid cohesion protein 112, sister chromatid cohesion protein PDS5 homolog A | |
| Produced in rabbits immunized with E. coli-derived Human SCC112 fragment. | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair | |
| 23244 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction