missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | SCD |
|---|---|
| Concentration | 400 μg/mL |
| Applications | Immunofluorescence, Immunohistochemistry, Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17761079
|
Proteintech
28678-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17751079
|
Proteintech
28678-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SCD Polyclonal antibody specifically detects SCD in Human samples. It is validated for IF, IHC, WB, ELISASpecifications
| SCD | |
| Immunofluorescence, Immunohistochemistry, Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| IgG | |
| Antigen affinity purification | |
| Antibody |
| 400 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human | |
| 6319 | |
| Primary | |
| Store at -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title