Learn More
DWK Life Sciences Screw Cap closed, with seal for safety standard ground sockets
Shop All DWK Life Sciences ProductsDescription
With closed screw caps, devices made of DURAN™ safety grinding can be securely closed after use. The PTFE-coated silicone seal provides maximum chemical resistance.
- Secure closure of the safety ground joints prevents unintentional opening of the ground joint
- PTFE-coated silicone gasket for maximum chemical resistance
Specifications
Specifications
| Quantity | 10/Pk. |
| Item Description | Suitable for sealing security grinding |
| Joint Size | NS 19/26 |