missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCRN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11485-2-AP-20UL
This item is not returnable.
View return policy
Description
SCRN2 is a member of the secernin family of proteins which includes the homologous SCRN1, a protein that was first identified as being involved in the regulation of exocytosis from peritoneal mast cells. Little is known of SCRN2, but studies have shown that SCRN1 may possess epitopes that function as tumor-associated antigens in gastric cancers and increased SCRN1 expression in patients with colorectal cancer correlated with poor prognosis, suggesting that SCRN2 may also be involved in protein secretion or the regulation of cell growth.Specifications
| SCRN2 | |
| Polyclonal | |
| Unconjugated | |
| SCRN2 | |
| SCRN2, secernin 2, Ses2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 217140, 360612, 90507 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6AYR8, Q8VCA8, Q96FV2 | |
| Scrn2 | |
| SCRN2 Fusion Protein Ag2001 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction