missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHB Antibody (CL0347) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52920JF646
This item is not returnable.
View return policy
Description
SDHB Monoclonal antibody specifically detects SDHB in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| SDHB | |
| Monoclonal | |
| Janelia Fluor 646 | |
| EC 1.3.5.1, FLJ92337, IP, Iron-sulfur subunit of complex II, PGL4, SDH, SDH1, SDH2, SDHIP, succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial, succinate dehydrogenase complex, subunit B, iron sulfur (Ip) | |
| This antibody was developed using a recombinant protein derived from P21912, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer | |
| 6390 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL0347 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction