Learn More
SDHB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10620-1-AP-150UL
 View more versions of this product
Product Code. 16827113
  • 410.00€ / 150µL
Description
Complex II of the respiratory chain, which is specifically involved in the oxidation of succinate, carries electrons from FADH to CoQ. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. The iron-sulfur subunit is highly conserved and contains three cysteine-rich clusters which may comprise the iron-sulfur centers of the enzyme. Sporadic and familial mutations in this gene result in paragangliomas and pheochromocytoma, and support a link between mitochondrial dysfunction and tumorigenesis.
Specifications
Specifications
SDHB
Polyclonal
Unconjugated
SDHB
IP, PGL4, SDH, SDH1, SDHB, SDHIP
Rabbit
Antigen Affinity Chromatography
RUO
298596, 6390, 67680
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P21912, P21913, Q9CQA3
SDHB
SDHB Fusion Protein Ag0968
150 μL
Primary
Human, Mouse, Rat