missing translation for 'onlineSavingsMsg'
Learn More

Sea Sand, Acid Washed and Calcinated For Laboratory Use and DAB, Chem Lab

Product Code. 16234291
Change view
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16234291 5 kg 5kg
15949296 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16234291 Supplier Chem-Lab Supplier No. CL00.2631.5000
  • ONLINE EXCLUSIVE PRICE
    166.00€
    129.00€ / 5kg
    Save 37.00€
    22% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Color Gray
Physical Form Solid
Quantity 5 kg
Grade Analytical
Loss on Ignition < 0.05%
Packaging Polyethylene
Density 2.66 g/cm3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.