missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sebuthylazine-d5 (ethyl-d5), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Sebuthylazine D5 |
| CAS | 1219805-56-7 |
| Molecular Formula | C9 2H5 H11 Cl N5 |
| Quantity | 0.01 g |
| Synonym | Sebuthylazine D5 (N-ethyl D5), 6-Chloro-N4-(1,1,2,2,2-pentadeuterioethyl)-N2-sec-butyl-1,3,5-triazine-2,4-diamine |
| InChI Formula | InChI=1 S/C9H16ClN5/c1-4-6(3)12-9-14-7(10)13-8(15-9)11-5-2/h6H,4-5H2,1-3H3,(H2,11,12,13,14,15)/i2D3,5D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])Nc1nc(Cl)nc(NC(C)CC)n1 |
| IUPAC Name | 2-N-butan-2-yl-6-chloro-4-N-(1,1,2,2,2-pentadeuterioethyl)-1,3,5-triazine-2,4-diamine |
| Molecular Weight (g/mol) | 234.74 |
| Formula Weight | 234.1408 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction