missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEC22B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14776-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine. The encoded protein is involved in myelopoiesis, and defects in this gene have been associated with several myeloproliferative disorders. Two variants encoding different isoforms have been found for this gene.Specifications
| SEC22B | |
| Polyclonal | |
| Unconjugated | |
| SEC22B | |
| ER Golgi SNARE of 24 kDa, ERS 24, ERS24, SEC22B, SEC22L1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20333, 310710, 9554 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08547, O75396, Q4KM74 | |
| Sec22b | |
| SEC22B Fusion Protein Ag6077 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction