Learn More
WLD Tec™ Security Adapter
Product Code. 10487151
Change view
Click to view available options
Length (Metric):
0.5 m
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10487151 0.5 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10487151 Supplier WLD Tec™ Supplier No. 6.000.800
  • ONLINE EXCLUSIVE PRICE
    174.30€
    142.00€ / Pack of 1
    Save 32.30€
    18% Off
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Product Type Gas Adapter
Length (Metric) 0.5 m
Includes DVGW Tubing (0.5 m) for Cartridges C 206, 190 g from Camping Gaz, Integrated Pressure Regulator 50 mbar, Shut-Off Valve