Learn More
ERLAB Segmented Worksurface
Product Code. 30484794 Shop All Erlab Products
Change view
Click to view available options
For Use With (Equipment):
SOLIS 1200
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
30484794 SOLIS 1200 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30484794 Supplier ERLAB Supplier No. NORT000480
  • ONLINE EXCLUSIVE PRICE
    106.00€
    98.00€ / Pack of 1
    Save 8.00€
    7% Off
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Enhances flexibility and ease of maintenance in safety cabinets, designed for modular use
  • The Segmented Worksurface enhances flexibility
  • Ease of maintenance in safety cabinets
  • Designed for modular use
  • Allows partial removal for cleaning or setup changes
Specifications
Product Type Workstation
Housing Material Segmented Stainless Steel
For Use With (Equipment) SOLIS 1200