Description
- Available as either Cy3 or Cy5
- Example protocols included with each dye pack
Specifications
Specifications
| For Use With (Application) | Steady state Fluorescence, FP and FRET |
| Product Type | Cy5 |
| Quantity | 3 x 0.5 mg |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1