Learn More
- Home
- Products
- Sterilizers and Autoclaves
- Autoclave Bags, Wraps, and Sleeves
- Keystone Cleanroom Products Self Seal Tyvek™ Steam Sterilization Pouches
Keystone Cleanroom Products Self Seal Tyvek™ Steam Sterilization Pouches
Product Code. 17750368
Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
3.5 x 8 in.
35.25 x 16 in.
6 x 2 in.
6 x 3 in.
8 x 3 in.
8 x 3.5 in.
8 x 6.125 in.
9 x 2.5 in.
10 x 5 in.
10 x 8 in.
12 x 6 in.
13 x 7 in.
15 x 4 in.
15 x 8.125 in.
18 x 8 in.
22 x 4 in.
28 x 6 in.
32 x 6 in.
34 x 8 in.
35 x 6 in.
4 x 15 in.
5 x 10 in.
6 x 35 in.
8 x 10 in.
8 x 15 in.
8 x 34 in.
10 x 11 in.
10 x 20 in.
12 x 10 in.
14 x 12 in.
15 x 20 in.
16 x 26 in.
16 x 32 in.
17 x 12 in.
17 x 16 in.
20 x 10.125 in.
20 x 15 in.
22 x 18 in.
24 x 14 in.
26 x 16 in.
26 x 24 in.
30 x 24 in.
32 x 16 in.
35 x 24 in.
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
44 product options available for selection
| Product Code. | Dimensions (L x W) | Unit Size |
|---|---|---|
| 17750368 | 22 x 18 in. | Pack of 500 |
| 17841701 | 3.5 x 8 in. | Pack of 500 |
| 17851701 | 4 x 15 in. | Pack of 500 |
| 17861701 | 5 x 10 in. | Pack of 500 |
| 17707827 | 6 x 2 in. | Pack of 1000 |
| 17727827 | 6 x 3 in. | Pack of 1000 |
| 17861711 | 6 x 35 in. | Pack of 250 |
| 17737827 | 8 x 3 in. | Pack of 500 |
| 17747827 | 8 x 3.5 in. | Pack of 500 |
| 17787827 | 8 x 6.125 in. | Pack of 500 |
| 17871701 | 8 x 10 in. | Pack of 500 |
| 17881701 | 8 x 15 in. | Pack of 500 |
| 17891701 | 8 x 34 in. | Pack of 500 |
| 17717827 | 9 x 2.5 in. | Pack of 1000 |
| 17777827 | 10 x 5 in. | Pack of 500 |
| 17747837 | 10 x 8 in. | Pack of 500 |
| 17801711 | 10 x 11 in. | Pack of 500 |
| 17811711 | 10 x 20 in. | Pack of 500 |
| 17797827 | 12 x 6 in. | Pack of 500 |
| 17787837 | 12 x 10 in. | Pack of 500 |
| 17737837 | 13 x 7 in. | Pack of 500 |
| 17707847 | 14 x 12 in. | Pack of 500 |
| 17757827 | 15 x 4 in. | Pack of 500 |
| 17757837 | 15 x 8.125 in. | Pack of 500 |
| 17831711 | 15 x 20 in. | Pack of 500 |
| 17841711 | 16 x 26 in. | Pack of 500 |
| 17851711 | 16 x 32 in. | Pack of 250 |
| 17717847 | 17 x 12 in. | Pack of 500 |
| 17747847 | 17 x 16 in. | Pack of 500 |
| 17767837 | 18 x 8 in. | Pack of 500 |
| 17797837 | 20 x 10.125 in. | Pack of 500 |
| 17737847 | 20 x 15 in. | Pack of 500 |
| 17767827 | 22 x 4 in. | Pack of 500 |
| 17727847 | 24 x 14 in. | Pack of 500 |
| 17757847 | 26 x 16 in. | Pack of 500 |
| 17777847 | 26 x 24 in. | Pack of 250 |
| 17707837 | 28 x 6 in. | Pack of 500 |
| 17760368 | 30 x 24 in. | Pack of 250 |
| 17717837 | 32 x 6 in. | Pack of 500 |
| 17740368 | 32 x 16 in. | Pack of 250 |
| 17777837 | 34 x 8 in. | Pack of 500 |
| 17727837 | 35 x 6 in. | Pack of 500 |
| 17787847 | 35 x 24 in. | Pack of 250 |
| 17767847 | 35.25 x 16 in. | Pack of 250 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 44 options available.
Description
- Great choice for equipment parts that are heavy, have sharp edges, or packaging that requires surface sanitization post-sterilization
- DuPont Tyvek™/Polyester-based flexible film is lint-free, moisture and puncture resistant, extremely tough, and designed to be autoclaved
- The Tyvek™ side offers high breathability while providing an excellent barrier from microbial contamination
- The clear film side offers excellent clarity along with strength and durability
- These bags feature a medical grade adhesive strip for convenient self-sealing
- Each pouch has a CLASS A process indicator, which changes from pink to dark brown when exposed to steam
- When the peel-able seals are opened, virtually no airborne particulates are generated
- Recommended for sterilization of pharmaceutical equipment
- Manufactured and packaged in controlled cleanroom environment
- Eliminates the need for the use and validation of a Heat Sealer
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance (COC) |
| Closure Type | Self-seal |
| Color | White |
| Disposable | Single-use |
| For Use With (Application) | Steam Sterilization |
| Material | Tyvek |
| Sterility | Non-sterile |
| Latex Free | Yes |
| Quantity | 500/Pk. |
| Dimensions (L x W) | 22 x 18 in. |
| Length (English) | 22 in. |
| Width (English) | 18 in. |
| Product Type | Autoclave Bag |
| Length (Metric) | 55.9 cm |
| Width (Metric) | 45.7 cm |
| Show More Show Less |