Learn More
Globe Scientific Self-Standing Transport Tubes
Product Code. 31922703 Shop All Globe Scientific Products
Change view
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31922703 10 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31922703 Supplier Globe Scientific Supplier No. 6102B
Estimated Shipment: 04-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Globe Scientific Inc. Self-Standing Transport Tubes with leak-resistant screw caps
  • Designed for the safe storage and transport of biological materials
  • Transparent polypropylene (PP) vials allow unobstructed viewing of the sample
  • Vials are shatter-proof and may be exposed to freezing temperatures
  • Leak-resistant screw caps can be removed or sealed with a single turn
Specifications
Color Natural
Closure Color Blue
Autoclavable Autoclavable
Capacity (Metric) 10 mL
Bottom Shape Conical (Self-standing)
Material Polypropylene
Style Skirt Base
Sterility Non-sterile
Graduated Yes
Closure Material Polyethylene
Height (Metric) 92 mm
Closure Type Screw Cap
Product Type Transport Tube (Self-Standing)