missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA4B Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149163
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Inhibits axonal extension by providing local signals to specify territories inaccessible for growing axons. [UniProt].
Specifications
| SEMA4B | |
| Polyclonal | |
| Unconjugated | |
| SEMA4B | |
| KIAA1745; mKIAA1745; sema C; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4B; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4B; Sema4b; SEMAC; semaphorin 4B; semaphorin-4B; Semaphorin-C; SemC; UNQ749/PRO1480 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 20352 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q62179 | |
| SEMA4B | |
| Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 4B, Leu31-Glu703. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction