missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA6A Monoclonal Antibody (169203)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA560928
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
SEMA6A can act as repulsive axon guidance cues. It may play a role in channeling sympathetic axons into the sympathetic chains and controlling the temporal sequence of sympathetic target innervation.
Specifications
| SEMA6A | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9H2E6 | |
| SEMA6A | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6A, Gly19-Thr649. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 169203 | |
| Unconjugated | |
| SEMA6A | |
| 9330158E07; A730020P05Rik; AI851735; b2b997Clo; HT018; KIAA1368; Mutant line 997; SEMA; sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A; sema Q; Sema VIA; SEMA6A; SEMA6A1; SEMA6A-1; semaphorin 6A; semaphorin 6A-1; semaphorin Q; Semaphorin VIA; semaphorin-6A; Semaphorin-6A-1; Semaq; VIA | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 57556 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction