missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaphorin 4D/CD100 Antibody (547412), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB52351Y
This item is not returnable.
View return policy
Description
Semaphorin 4D/CD100 Monoclonal antibody specifically detects Semaphorin 4D/CD100 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Semaphorin 4D/CD100 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 547412 | |
| Flow Cytometry, CyTOF-ready | |
| A8, CD100 antigen, CD100M-sema-G, coll-4, FLJ33485, FLJ34282, FLJ39737, FLJ46484, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, 4D, MGC169138, MGC169141, sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and shortcytoplasmic domain, (semaphorin) 4D, semaphorin-4D | |
| Mouse myeloma cell line NS0-derived recombinant mouse Semaphorin 4D/CD100, Phe24-Thr657, Accession # NP_038688 | |
| 0.1 mL | |
| Apoptosis, Immunology | |
| 10507 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction