Learn More
Semperit™ Semperguard™ Nitrile Comfort Powder-free Gloves
Product Code. p-8376028
Change view
Click to view available options
Size:
7/8
8/9
9/10
Small
Unit Size:
Pack of 100
Pack of 90
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15564456 Small Pack of 100
15574456 7/8 Pack of 100
15584456 8/9 Pack of 100
15594456 9/10 Pack of 90
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15564456 Supplier Semperit™ Supplier No. 3000001643
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Disposable protective glove made of Nitrile. Semperit™ Semperguard™ Nitrile Comfort Powder-free Gloves are latex free with high wearing comfort.
  • CE Category III protective glove for complex risks. CE Class I examination glove.
  • Special nitrile formulation with optimized resistance to oils and fats, good tear-resistance
  • Excellent wearing comfort
  • Tested and certified according to 1935/2004 - suitable for food contact
  • The problem-solver for single use to protect hands against chemical splashing and in the processing of fatty foods.
  • Free from phthalates / softeners and allergenic latex proteins
  • Signal color blue – suitable for areas subject to HACCP standards
Specifications
Color Blue
Material Nitrile
Powdered Powder Free
Size Small
Thickness (English) 7.8 mil
Thickness (Metric) 0.2 mm
Glove Cuff Style Rolled
Model 3000004106
Length (Metric) 240 mm