missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sensititre AIM™ Automated Inoculation Delivery System

Product Code. 15673434
Change view
Click to view available options
Height (English):
11.3 in.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (English) Unit Size
15673434 11.3 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 15673434 Supplier Thermo Scientific™ Supplier No. YV3020
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.

Quickly and accurately dose most 96-well microtitre plates, eliminating both skipped wells and costly repeat tests.

Quickly and accurately dose most 96-well microtitre plates, eliminating both skipped wells and costly repeat tests, with the Thermo Scientific™ Sensititre AIM™ Automated Inoculation Delivery System. Just select your dosing volume and pattern with the easy-to-use, icon-driven touch screen for simple, intuitive plate inoculation. With its compact, user-friendly design, the AIM System also provides flexible storage and accessibility with minimal benchtop footprint to conserve valuable laboratory space.

  • Compatibility with most 96-well microtitre plates ensures flexibility with dry or frozen susceptibility formats
  • Unique tube design and manually applied seal provide additional protection against biohazardous spill concerns
Not all products are available for sale in all territories. Please inquire.

VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Description Sensititre AIM Automated Inoculation Delivery System
Product Type Microbiology Media
Test Type Minimum Inhibitory Concentration (MIC)
Regulatory Status FDA-cleared
Width (English) 13.3 in.
Width (Metric) 33.8 cm
Height (English) 11.3 in.
Height (Metric) 28.7 cm
Depth (English) 12.3 in.
Depth (Metric) 31.2 cm
Weight (English) 17.6 lb.
Weight (Metric) 8 kg
Electrical Requirements 94-237V; 47-63Hz; 150w
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.