missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sensititre™ Avian AVIAN1F Vet AST Plate

Product Code. 17287260
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17287260 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17287260 Supplier Thermo Scientific™ Supplier No. YAVIAN1F
  • ONLINE EXCLUSIVE PRICE
    325.00€
    190.17€ / Pack of 10
    Save 134.83€
    41% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform accurate AST with an avian- and poultry-specific formulary using this convenient and cost-effective plate.

Perform accurate AST with an avian- and poultry-specific formulary using Thermo Scientific™ Sensititre™ Avian AVIAN1F AST Plate. Sensititre Avian AVIAN1F AST Plate delivers accurate results on a single plate format reducing the need for offline tests and retests.

Achieve reliable, consistent results for non-fastidious Gram-positive and Gram-negative isolates of veterinary origin with Sensititre Avian AVIAN1F AST Plate.

  • Extended shelf life, room temperature storage eliminates inventory control concerns.
  • Individual packaging allows laboratory to test one plate at a time with no waste.
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology.
  • Automated or manual read options are available for ultimate flexibility.

Not all products are available for sale in all territories. Please inquire.
 

VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Antimicrobial Agents Various
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Non-fastidious, Gram-positive and Gram-negative Veterinary Isolates
Test Type Veterinary AST
Isolates Per Plate 1
Quantity 10/Pk.
Reading Options Autoread or manual
Specimen Type One-Isolate, Avian
Species Avian
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.