Learn More
Thermo Scientific™ Sensititre™ EU Surveillance ESBL EUVSEC2 AST Plate
Product Code. 15635938
Change view
Click to view available options
Quantity:
10 Plates/Box
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15635938 10 Plates/Box Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15635938 Supplier Thermo Scientific™ Supplier No. YEUVSEC2
  • ONLINE EXCLUSIVE PRICE
    323.00€
    137.27€ / Pack of 10
    Save 185.73€
    57% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Comply with EU Surveillance System program for ESBL isolates with this convenient plate.
Perform accurate AST with ESBL isolates specific formulary as a part of EU Surveillance program with Thermo Scientific™ Sensititre EU Surveillance ESBL EUVSEC2 AST Plate.
Comply with EU Surveillance System program for ESBL isolates with Sensititre EU Surveillance EU Surveillance ESBL EUVSEC2 AST Plate.
  • Extended shelf life, room temperature storage eliminates inventory control concerns
  • Individual packaging allows a laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • Automated or manual read options are available for ultimate flexibility
Antimicrobial Resistance (AMR) is a significant concern and of increasing importance. The inappropriate use of antimicrobial agents in humans and animals is leading this growing and dangerous issue, emerging as a public health threat. This is why the European Parliament, the Council and its Agencies: European Centre for Disease Prevention and Control (ECDC), the European Food Safety Authority (EFSA) and the European Medicines Agency (EMA), have identified the need to establish a common strategy in the fight against AMR. 
The Sensititre System utilizes true minimum inhibitory concentration (MIC) results, which are essential to identifying antimicrobial resistance, and provide greater sensitivity for better resistance tracking.
With a wide range of Sensititre plates specifically designed for surveillance programs, the Sensititre System is a leader in antimicrobial susceptibility testing for surveillance.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand
TRUSTED_SUSTAINABILITY
Specifications
Description Sensititre EU Surveillance ESBL EUVSEC2 AST Plate
Product Type AST Plate
Test Type Antimicrobial Surveillance, Minimum Inhibitory Concentration (MIC)
Isolates Per Plate 1
No. per Pack 10
Packaging Type Box
Quantity 10 Plates/Box
Reading Options Manual, Vizion
Regulatory Status RUO
Specimen Type ESBL Isolates
Species ESBL/AmpC/carbapenemase-producing Escherichia coli