Learn More
Sensititre™ Gram Negative GNX3F AST Plate
Product Code. 17779217
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17779217 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17779217 Supplier Thermo Scientific Oxoid Supplier No. YGNX3F
  • ONLINE EXCLUSIVE PRICE
    319.00€
    148.61€ / Pack of 10
    Save 170.39€
    53% Off
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Perform susceptibility testing of Gram-negative isolates with this research use only plate containing Colistin, Doripenem, Doxycycline and Polymyxin B
Perform susceptibility testing of non-fastidious Gram-negative isolates on a single platform with the Thermo Scientific™ Sensititre™ Gram Negative GNX3F AST Plate. The Sensititre™ Gram Negative GNX3F AST Plate is intended for research use only.
Decrease costs associated with multiple test methods with 21 antimicrobial compounds on a single plate, including Colistin and Polymyxin B.
Test most bacteria, including challenging resistant organisms, on a single platform with the Sensititre Gram Negative GNX3F AST Plate Plate.
  • Automated or manual read options are available for ultimate flexibility
  • Extended shelf life and room-temperature storage simplify inventory control
  • Individual packaging allows laboratories to test one plate at a time with no waste.
Not all products are available for sale in all territories. Please inquire. 
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
0.5 McFarland Standard H2O (Part No. T3339)
Antimicrobial Agents Various
Broth Type MHB (Part No. T3462)
Description SensititreGram Negative Research Use Only MIC Plate for Non-Fermenters
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Non-fastidious, Gram-negative Organisms
Test Type Minimum Inhibitory Concentration (MIC)
Incubation Conditions 35°C O2
Incubation Time 18 hr
Isolates Per Plate 1
No. per Pack 10
Packaging Type Box
Quantity 10/Pk.
Reading Options Autoread or manual
Transfer Volume For Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL
Volume Per Well 50 μL