Learn More
Thermo Scientific™ Sensititre™ Gram Positive MIC Plate
Product Code. 17847144
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17847144 10/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17847144 Supplier Thermo Scientific™ Supplier No. YGPN3F
  • ONLINE EXCLUSIVE PRICE
    382.00€
    160.70€ / Pack of 1
    Save 221.30€
    57% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Monitor resistance with a complete selection of antimicrobials using the Thermo Scientific™ Sensititre™ Gram Positive MIC Plate.
Monitor resistance with a complete selection of antimicrobials using the Sensititre Gram Positive MIC Plate.
  • Plate has fewest limitations for Enterococcus spp. and Staphylococcus spp. versus other automated systems
  • Full-range, true MIC readings provide results for better detection of resistance
  • Automated or manual read options are available for ultimate flexibility
  • MIC plate contains antimicrobic only; broth added at time of test extends shelf life and ensures accurate, easy-to-read results for Gram positive isolates
  • Extended 12 to 24 month shelf life and room-temperature storage simplify inventory control
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
0.5 McFarland Standard H2O (Part No. T3339)
Antimicrobial Agents Various
Broth Type MHB (Part No. T3462)
Description Sensititre Gram Positive MIC Plate
Antibiotic Code Various
Product Type AST Plate
Target Organism Class Gram-positive Organisms
Test Type Minimum Inhibitory Concentration (MIC)
Incubation Conditions 35°C O2
Incubation Time 18-24 hr
Isolates Per Plate 1
No. per Pack 10
Packaging Type Box
Quantity 10/Pk.
Reading Options Autoread or manual
Transfer Volume 10µL or 30µL* (*Aid in detection of resistance mechanisms for gram positive organisms; see pack insert)
Volume Per Well 50 μL