Learn More
Thermo Scientific™ Sensititre™ NARMS Gram Negative Plate
Description
Perform accurate AST with a Gram negative isolate-specific formulary with the Thermo Scientific™ Sensititre™ NARMS Gram Negative Plate.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Antimicrobial Agents | Various |
| Description | Sensititre NARMS Gram Negative Plate |
| Antibiotic Code | Various |
| Product Type | AST Plate |
| Target Organism Class | Gram-negative Microorganisms |
| Test Type | Microbial Susceptibility |
| Isolates Per Plate | 1 |
| Quantity | 10/Pk. |
| Reading Options | Autoread or manual |