missing translation for 'onlineSavingsMsg'
Learn More

WTW™ SenTix™ Special pH Electrodes
GREENER_CHOICE

Product Code. 10663742
Change view
Click to view available options
Connector Types:
Fixed Cable
S7 Plug Head
Temperature (Metric) Range:
-5°C to +100°C
0°C to 60°C
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types Temperature (Metric) Range unitSize
10663742 S7 Plug Head 0°C to 60°C Each
15153976 Fixed Cable -5°C to +100°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10663742 Supplier WTW™ Supplier No. 103650
  • ONLINE EXCLUSIVE PRICE
    341.22€
    314.00€ / Pack of 1
    Save 27.22€
    7% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Specialists for all cases - SenTix™ pH Special Electrodes for special applications can determine the pH value of surfaces, suspended solids, suspensions and much more.

  • Adaptation to the sample consistency by special glass membranes
  • Optimal measuring accuracy by adapted diaphragm technique
  • Available for either low-conducting samples or for emulsions
TRUSTED_SUSTAINABILITY

Specifications

Description For low-conducting samples, S7 plug head
Connector Types S7 Plug Head
Material Glass Shaft, Cut Diaphragm
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 60°C
Type SenTix™ PLUS Special pH Electrode
For Use With (Equipment) Vario pH meter
Includes DIN, BNC electrode plug
Model SenTix™ HW
Temperature (Metric) Range 0°C to 60°C
Length (Metric) Stem 170 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.