missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPT8 Mouse anti-Human, Clone: 4A12G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66188-1-IG-150UL
This item is not returnable.
View return policy
Description
SEPT8 is a member of the highly conserved septin family. Septins are 40- to 60-kDa GTPases that assemble as filamentous scaffolds. They are involved in the organization of submembranous structures, in neuronal polarity, and in vesicle trafficking (Blaser et al., 2003 ).Specifications
| SEPT8 | |
| Monoclonal | |
| 0.71 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q92599 | |
| SEPT8 | |
| SEPT8 Fusion Protein Ag19114 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 4A12G9 | |
| Unconjugated | |
| SEPT8 | |
| KIAA0202, SEP2, SEPT8, septin 8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23176 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction