Learn More
Applied Biosystems™ Sequenase Version 2.0 DNA Sequencing Kit
Product Code. 15899886
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. Quantity unitSize
15899886 1 Kit Each
1 options
Product Code. 15899886
Brand: Applied Biosystems™ 707701KT
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Includes
Sequenase Version 2.0 DNA Polymerase, Inorganic Pyrophosphatase, Enzyme Dilution Buffer, Glycerol Enzyme Dilution Buffer, Sequenase Reaction Buffer (5X), Dithiothreitol Solution, Mn Buffer, Control DNA M13 mp18, Primer (-40 Universal), Labeling Mix (dGTP, 5X), ddGTP Termination Mix (for dGTP), ddATP Termination Mix (for dGTP), ddTTP Termination Mix (for dGTP), ddCTP Termination Mix (for dGTP), Sequencing Extending Mix (for dGTP), Labeling Mix (dITP, 5X), ddGTP Termination Mix (for dITP), ddATP Termination Mix (for dITP), ddTTP Termination Mix (for dITP), ddCTP Termination Mix (for dITP), Sequence Extending Mix (for dITP), Stop Solution, Protocol Book
The Sequenase™ Version 2.0 DNA Sequencing Kit features Sequenase Version 2.0 DNA Polymerase, the standard for high quality manual DNA sequencing.
TRUSTED_SUSTAINABILITY
Specifications
Format Kit
For Use With (Application) Sequencing
Polymerase Genetically-Engineered form of T7 DNA Polymerase
Product Type Sequencing Kit
Quantity 1 Kit