Learn More
Cytiva Sera-Mag SpeedBeads Amine-Blocked Particles
Product Code. 11854962 Shop All Cytiva Products
Change view
Click to view available options
Packaging:
1 mL
100 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging unitSize
11854962 1 mL Each
11844962 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11854962 Supplier Cytiva Supplier No. 19152104011150
Sera-Mag SpeedBeads Amine-Blocked Particles reduce undesired protein adsorption.
  • Combine a fast magnetic response time and high binding capacity with a large surface area, high sensitivity, stability, physical integrity, and fast reaction kinetics
  • Typical applications include molecular biology, nucleic acid isolation, research, and clinical diagnostic immunoassays
  • The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer
  • Two layers of magnetite are coated onto this core, resulting in faster magnetic response times
  • The surface is blocked with a non-surfactant, non-protein based method, to help prevent non-specific binding of proteins
  • TRUSTED_SUSTAINABILITY
    Specifications
    Binding Property Amine
    Concentration 10 mg/mL
    Material Magnetite
    Packaging 1 mL
    Diameter (Metric) 1 μm