Learn More
Cytiva Sera-Mag SpeedBeads Streptavidin-Blocked Magnetic Particles
Product Code. 11814982 Shop All Cytiva Products
Change view
Click to view available options
Packaging:
1 mL
5 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging unitSize
11814982 1 mL Each
11894972 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11814982 Supplier Cytiva Supplier No. 21152104011150
Combines high affinity for biotinylated target molecules with very low nonspecific binding.
  • Combines fast reaction kinetics with very low non-specific binding for increased throughput and precision
  • Applications include automated immunoassays, immunoprecipitation, and protein purification
  • Streptavidin coating targets Biotinylated Molecules
  • Compounds that are difficult to attach to particle surfaces by conventional means may be amenable to biotinylation
  • Provides a high biotin-binding capacity along with a strong affinity for targeted, biotin labeled molecules
  • The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer
  • Two layers of magnetite are coated onto this core, resulting in faster magnetic response times
  • The surface is blocked with a non-surfactant, non-protein based method, to help prevent non-specific binding of proteins
  • TRUSTED_SUSTAINABILITY
    Specifications
    Concentration 10 mg/mL
    Packaging 1 mL
    Diameter (Metric) 1 μm