missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINB3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26558-1-AP-150UL
This item is not returnable.
View return policy
Description
SPB3 may act as a protease inhibitor to modulate the host immune response against tumor cells.Specifications
| SERPINB3 | |
| Polyclonal | |
| Unconjugated | |
| SERPINB3 | |
| HsT1196, Protein T4 A, SCC, SCCA, SCCA 1, SCCA PD, SCCA1, Serpin B3, SERPINB3, T4 A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6317 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P29508 | |
| SERPINB3 | |
| SERPINB3 Fusion Protein Ag24846 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction