missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Amyloid A1 Antibody (SAA1cc), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
553.00€
Specifications
| Antigen | Serum Amyloid A1 |
|---|---|
| Clone | SAA1cc |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Janelia Fluor 646 |
Description
Serum Amyloid A1 Monoclonal antibody specifically detects Serum Amyloid A1 in Human, Canine samples. It is validated for Western Blot, ELISASpecifications
| Serum Amyloid A1 | |
| Western Blot | |
| Janelia Fluor 646 | |
| Mouse | |
| Alzheimers Research, Biologically Active Proteins, Cancer, Neuroscience | |
| 50mM Sodium Borate | |
| 6288 | |
| IgG1 | |
| Protein A purified |
| SAA1cc | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human SAA. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title