Learn More
Abeba™ Service 3030 Shoes
A walking sole with cushioning effect, a comfortable footbed and finest upper leather guarantee the highest wearing comfort and a good well being until the end of work.
Brand:  Abeba™ 3030/36
Additional Details : Weight : 0.82000kg
 View more versions of this product
Product Code. 15618267
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • CE EN ISO 20347:2012 O1 FO SRC
  • Smooth leather
  • Black
  • Slip resistance outsole sole
  • Integrated footbed
  • Lining microfiber, respiration absorbing
  • Adjustable instep area
  • velcro fastening
Specifications
Specifications
CE EN ISO 20347:2012 O1 FO SRC
Microfiber
36
3030
Black
Leather
Slip-resistant