missing translation for 'onlineSavingsMsg'
Learn More

SET1B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

220.00€ - 326.00€

Specifications

Antigen SET1B
Dilution Immunocytochemistry/ Immunofluorescence 1:100-1:500
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18675508
View Documents
Novus Biologicals
NBP3-06532-100ul
100 μg
326.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18617837
View Documents
Novus Biologicals
NBP3-06532-50ul
50 μg
220.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SET1B Polyclonal antibody specifically detects SET1B in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

SET1B
Immunofluorescence
Unconjugated
Rabbit
Human
EC 2.1.1, FLJ20803, histone-lysine N-methyltransferase SETD1B, hSET1B, KIAA1076EC 2.1.1.43, KMT2GSET1B, Lysine N-methyltransferase 2G, SET domain containing 1B, SET domain-containing protein 1B, Set1B
Produced in rabbits immunized with E. coli-derived Human SET1B fragment.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunocytochemistry/ Immunofluorescence 1:100-1:500
Polyclonal
Purified
RUO
PBS, pH7.0
23067
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.