missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SETD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569594
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This function of this gene has yet to be determined but mutations in this gene have been associated with autosomal dominant mental retardation-23. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| SETD5 | |
| Polyclonal | |
| Unconjugated | |
| SETD5 | |
| 2900045N06Rik; C330007C20; C85544; Histone-lysine N-methyltransferase SETD5; histone-lysine N-methyltransferase SETD5; SET domain-containing protein 5; Kiaa1757; LOW QUALITY PROTEIN: SET domain-containing protein 5; mKIAA1757; RGD1310433; SET domain containing 5; SET domain-containing protein 5; SETD5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 297514 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| SETD5 | |
| synthetic peptide corresponding to a region of rat SETD5. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction