missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SETD5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577197
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This function of this gene has yet to be determined but mutations in this gene have been associated with autosomal dominant mental retardation-23. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| SETD5 | |
| Polyclonal | |
| Unconjugated | |
| SETD5 | |
| 2900045N06Rik; C330007C20; C85544; Histone-lysine N-methyltransferase SETD5; histone-lysine N-methyltransferase SETD5; SET domain-containing protein 5; Kiaa1757; LOW QUALITY PROTEIN: SET domain-containing protein 5; mKIAA1757; RGD1310433; SET domain containing 5; SET domain-containing protein 5; SETD5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 297514, 55209, 72895 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5XJV7, Q9C0A6 | |
| SETD5 | |
| Recombinant full length Human SETD5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction