missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFPQ Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00€ - 593.00€
Specifications
| Antigen | SFPQ |
|---|---|
| Dilution | Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18695855
|
Novus Biologicals
NBP2-48876-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660817
|
Novus Biologicals
NBP2-48876 |
0.1 mL |
593.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SFPQ Polyclonal antibody specifically detects SFPQ in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDownSpecifications
| SFPQ | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS (pH 7.2), 40% Glycerol | |
| 6421 | |
| IgG | |
| Immunogen affinity purified |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockdown Validated | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| 100 kDa DNA-pairing protein, DNA-binding p52/p100 complex, 100 kDa subunit, hPOMp100, polypyrimidine tract binding protein associated, polypyrimidine tract-binding protein-associated splicing factor, Polypyrimidine tract-binding protein-associated-splicing factor, PSFPOMP100, PTB-associated splicing factor, PTB-associated-splicing factor, splicing factor proline/glutamine rich (polypyrimidine tract binding proteinassociated), splicing factor proline/glutamine rich (polypyrimidine tract-bindingprotein-associated), splicing factor proline/glutamine-rich, splicing factor, proline- and glutamine-rich | |
| This antibody was developed against a recombinant protein corresponding to amino acids: IGYEANPGVPPATMSGSMMGSDMRTERFGQGGAGPVGGQGPRGMGPGTPAGYGRGREEYEGPNK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title