Learn More
SFPQ Mouse anti-Human, Unconjugated, Clone: 1G4A5, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | SFPQ |
|---|---|
| Clone | 1G4A5 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16889575
|
Proteintech
67129-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16879575
|
Proteintech
67129-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SFPQ antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SFPQ. This antibody recognizes human, mouse, rat antigen. The SFPQ antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| SFPQ | |
| 500 μg/mL | |
| Monoclonal | |
| Mouse | |
| 6421 | |
| Primary |
| 1G4A5 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.