missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFRP1 Monoclonal Antibody (C5), Invitrogen™

Mouse Monoclonal Antibody
208.00€ - 453.00€
Specifications
| Antigen | SFRP1 |
|---|---|
| Clone | C5 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17604046
|
Invitrogen™
MA543584 |
20 μL |
208.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17694036
![]() |
Invitrogen™
MA543583 |
100 μL |
453.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Secreted frizzled-related protein 1 (SFRP1) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP1 and SFRP5 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP1 is expressed in several human tissues, with the highest levels in heart.Specifications
| SFRP1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| 2210415K03Rik; AW011917; AW107218; AW742929; frizzled-related protein; FRP; FRP1; FRP-1; FrzA; SARP2; SARP-2; Secreted apoptosis-related protein 2; secreted frizzled related protein 1; secreted frizzled-related protein 1; secreted frizzled-related sequence protein 1; SFRP1; sFRP-1 | |
| SFRP1 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| C5 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8N474 | |
| 6422 | |
| Ser32-Lys314 (Accession Q8N474), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SFRP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title