Learn More
Invitrogen™ SFT2D2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5101955
Product Code. 16698424
  • 493.00€ / 100µL
Description
Antibody detects endogenous levels of total SFT2B.
May be involved in fusion of retrograde transport vesicles derived from an endocytic compartment with the Golgi complex.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SFT2D2
Polyclonal
Unconjugated
SFT2D2
2010005O13Rik; AI506168; AW561909; cI-45; dJ747L4.C1.2; RGD1310623; SFT2 domain containing 2; SFT2 domain-containing protein 2; SFT2D2; sft2d2 {ECO:0000250; UniProtKB:O95562}; UNQ512; UNQ512/PRO1027; Vesicle transport protein SFT2B
Rabbit
Affinity Chromatography
RUO
108735, 360868, 375035
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O95562, Q4FZV2, Q8VD57
SFT2D2
A synthesized peptide derived from human SFT2D2(Accession O95562), corresponding to amino acid residues Y69-W119.