Learn More
Trajan™ HT8 GC Capillary Columns
Unique, high temperature phase designed for the analysis of polychlorinated biphenyl (PCB) congeners
Brand:  Trajan™ 054682
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15288693
  • 1565.00€ / Each
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • High temperature, low bleed column
  • Preferred column for polychlorinated biphenyl (PCB) analysis
  • Separates PCBs on ortho ring as well as boiling point
Specifications
Specifications
0.32 mm
8% Phenyl (equiv.) Polycarborane Siloxane
Polynuclear Aromatic Hydrocarbons Analysis, PCB Congener Analysis, Nitro-substituted Aromatics, Polynuclear Aromatic Hydrocarbons, Pesticides and Herbicides Analysis
0.25 μm
Forte™ HT8
End up with Pentavalent Bonds with Shared Sigma Bonds rather than piBond
360/370°C
-20°C to +360/370°C
60 m
Fused Silica