Learn More
Trajan™ HT8 GC Capillary Columns
Unique, high temperature phase designed for the analysis of polychlorinated biphenyl (PCB) congeners
820.00€ - 1565.00€
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
10157060
|
Trajan™
054675 |
0.22 mm | 25 m |
820.00€
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
15288693
|
Trajan™
054682 |
0.32 mm | 60 m |
1565.00€
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10701232
|
Trajan™
054680 |
0.32 mm | 25 m |
910.00€
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10257493
|
Trajan™
054677 |
0.25 mm | 30 m |
820.00€
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10432905
|
Trajan™
054683 |
0.25 mm | 60 m |
1375.00€
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
Description
- High temperature, low bleed column
- Preferred column for polychlorinated biphenyl (PCB) analysis
- Separates PCBs on ortho ring as well as boiling point
Specifications
360/370°C | |
0.25 μm |