Learn More
Trajan™ HT8 GC Capillary Columns
Unique, high temperature phase designed for the analysis of polychlorinated biphenyl (PCB) congeners
820.00€ - 1565.00€
Specifications
Max. Temperature 360/370°C
Film Thickness 0.25 μm
View More Specs
Products 5
Product Code Brand Diameter (Metric) Inner Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Length (Metric) Price Quantity & Availability  
10157060
View Documents Promotion Details
Trajan™
054675
0.22 mm 25 m
820.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
15288693
View Documents Promotion Details
Trajan™
054682
0.32 mm 60 m
1565.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
10701232
View Documents Promotion Details
Trajan™
054680
0.32 mm 25 m
910.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
10257493
View Documents Promotion Details
Trajan™
054677
0.25 mm 30 m
820.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
10432905
View Documents Promotion Details
Trajan™
054683
0.25 mm 60 m
1375.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • High temperature, low bleed column
  • Preferred column for polychlorinated biphenyl (PCB) analysis
  • Separates PCBs on ortho ring as well as boiling point
Specifications
Specifications
360/370°C
0.25 μm