Learn More
Trajan™ PEEKsil™ Tubing
Product Code. 15238753
Change view
Click to view available options
Diameter (Metric) Inner:
0.05 mm
0.175 mm
Diameter (Metric) Outer:
1.5 mm
1.6 mm
Overall Length (Metric):
50 mm
100 mm
200 mm
500 mm
Unit Size:
Pack of 2
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Diameter (Metric) Outer Overall Length (Metric) unitSize
15238753 0.05 mm 1.5 mm 50 mm Pack of 5
15298773 0.175 mm 1.5 mm 500 mm Pack of 2
15248753 0.05 mm 1.5 mm 100 mm Pack of 5
15208783 0.175 mm 1.5 mm 50 mm Pack of 5
15218783 0.175 mm 1.5 mm 100 mm Pack of 5
15228783 0.175 mm 1.5 mm 200 mm Pack of 5
15228753 0.05 mm 1.6 mm 500 mm Pack of 2
15258753 0.05 mm 1.5 mm 200 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15238753 Supplier Trajan™ Supplier No. 0624251
Only null left
Add to basket
Add to basket
Mechanically robust polymer-sheathed fused silica tubing. Trajan Scientific and Medical™ PEEKsil™ Tubing can be used for sealing with conventional metal or polymer ferrule systems.
  • Robust - PEEK polymer exterior and fused silica inner tubing
  • Smooth wall surface giving excellent flow characteristics
  • Compatible with most organic solvents
  • Inertness - extremely low absorption characteristics
  • Precise ID
Specifications
Type Tubing
Diameter (Metric) Outer 1.5 mm
Diameter (Metric) Inner 0.05 mm
Overall Length (Metric) 50 mm