Learn More
Trajan™ SolGel-WAX™ GC Capillary Columns
Comprised of Polyethylene Glycol in a Sol-Gel matarix. Trajan Scientific and Medical™ SolGel-WAX™ is a robust, high temperature polar WAX phase recommended fro highly active compounds.
Brand:  Trajan™ 054792
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 12935531
  • 1305.00€ / Each
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • Bonded polyethylene glycol encapsulated in synthetic glass (SolGel).
  • Ideal for mixtures containing alcohols, esters, aldehydes, ketones, or aromatics.
  • SolGel technology facilitates interactions between the sample and the phase, reduces column bleed and improves thermal stability.
  • Extremely stable and inert.
  • Columns are individually tested to ensure quality.
  • Replacement for all Wax columns, including acid or base stabilized.
Specifications
Specifications
0.32 mm
30°C to 260/280°C
60 m
0.5 μm
0.5 μm